Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 25 - 26 | 2633 | 108 | |
| GenBank mRNA | AK049419.1 | - | 24 - 25 | 2784 | 108 | |
| AK076357.1 | - | 24 - 25 | 3431 | 108 | ||
| AK152868.1 | - | 25 - 26 | 2621 | 108 | ||
| BC049929.1 | - | 13 - 14 | 1147 | 108 | ||
| BC079656.1 | - | 25 - 26 | 2633 | 6854039 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map