Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049419.1 | - | 3 - 4 | 502 | 87 | |
| AK076357.1 | - | 3 - 4 | 521 | 87 | ||
| AK152868.1 | - | 3 - 4 | 479 | 87 | ||
More Information
Set Membership: |
|

Ddx11
Genomic Map