Assay Details
| Gene Symbol: Tph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173391.3 | NP_775567.2 | 6 - 7 | 934 | 70 | |
| GenBank mRNA | AY090565.1 | - | 6 - 7 | 934 | 70 | |
| BC120514.1 | - | 6 - 7 | 925 | 40129437 | 70 | |
| BC125430.1 | - | 6 - 7 | 925 | 40129945 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tph2
Genomic Map