Assay Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177606.4 | NP_808274.2 | 14 - 15 | 2374 | 105 | |
| GenBank mRNA | AK029252.1 | - | 14 - 15 | 2376 | 105 | |
| AK173337.1 | - | 11 - 12 | 2029 | 105 | ||
| BC125583.1 | - | 14 - 15 | 2339 | 40130098 | 105 | |
| BC137804.1 | - | 14 - 15 | 2336 | 8860823 | 105 | |
More Information
Set Membership: |
|

Plekhh2
Genomic Map