Assay Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177606.4 | NP_808274.2 | 27 - 28 | 4143 | 75 | |
| GenBank mRNA | AK029252.1 | - | 27 - 28 | 4145 | 75 | |
| AK086794.1 | - | 7 - 8 | 973 | 75 | ||
| AK173337.1 | - | 24 - 25 | 3798 | 75 | ||
| BC125583.1 | - | 27 - 28 | 4108 | 40130098 | 75 | |
| BC137804.1 | - | 27 - 28 | 4105 | 8860823 | 75 | |
More Information
Set Membership: |
|

Plekhh2
Genomic Map