Assay Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177606.4 | NP_808274.2 | 5 - 6 | 498 | 137 | |
| GenBank mRNA | AK029252.1 | - | 5 - 6 | 500 | 137 | |
| AK173337.1 | - | 2 - 3 | 154 | 137 | ||
| BC021518.1 | - | 5 - 6 | 470 | 137 | ||
| BC125583.1 | - | 5 - 6 | 463 | 40130098 | 137 | |
| BC137804.1 | - | 5 - 6 | 460 | 8860823 | 137 | |
More Information
Set Membership: |
|

Plekhh2
Genomic Map