Assay Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177606.4 | NP_808274.2 | 9 - 10 | 1787 | 70 | |
| GenBank mRNA | AK029252.1 | - | 9 - 10 | 1789 | 70 | |
| AK173337.1 | - | 6 - 7 | 1443 | 70 | ||
| BC021518.1 | - | 9 - 10 | 1759 | 70 | ||
| BC125583.1 | - | 9 - 10 | 1752 | 40130098 | 70 | |
| BC137804.1 | - | 9 - 10 | 1749 | 8860823 | 70 | |
More Information
Set Membership: |
|

Plekhh2
Genomic Map