Assay Details
| Gene Symbol: Arr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133205.3 | NP_573468.1 | 3 - 4 | 144 | 68 | |
| GenBank mRNA | AF156979.1 | - | 3 - 4 | 85 | 68 | |
| BC017145.1 | - | 3 - 4 | 79 | 4506006 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF156979.1, BC017145.1, NM_133205.3 transcripts.
Set Membership: |
|

Arr3
Genomic Map