Assay Details
| Gene Symbol: Rmdn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201361.2 | NP_958749.1 | 2 - 3 | 549 | 74 | |
| GenBank mRNA | AK034617.1 | - | 2 - 3 | 551 | 74 | |
| AK139382.1 | - | 2 - 3 | 768 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rmdn2
Genomic Map