Assay Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183019.2 | NP_898840.2 | 2 - 3 | 463 | 75 | |
| GenBank mRNA | AK031310.1 | - | 2 - 3 | 446 | 75 | |
| AK034013.1 | - | 2 - 3 | 427 | 76 | ||
| AK134629.1 | - | 426 | 75 | |||
| AK138598.1 | - | 2 - 3 | 418 | 75 | ||
| AK163424.1 | - | 2 - 3 | 445 | 75 | ||
| AK169449.1 | - | 2 - 3 | 446 | 75 | ||
| AK169957.1 | - | 2 - 3 | 483 | 75 | ||
| BB609907.1 | - | 2 - 3 | 465 | 75 | ||
| BC055804.1 | - | 2 - 3 | 425 | 75 | ||
| BC094608.1 | - | 3983 | 75 | |||
| BC139371.1 | - | 2 - 3 | 446 | 8862393 | 75 | |
| BC139397.1 | - | 2 - 3 | 446 | 8862419 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef4
Genomic Map