Assay Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183019.2 | NP_898840.2 | 3 - 4 | 643 | 68 | |
| GenBank mRNA | AK031310.1 | - | 3 - 4 | 626 | 68 | |
| AK034013.1 | - | 3 - 4 | 607 | 68 | ||
| AK134629.1 | - | 606 | 68 | |||
| AK163424.1 | - | 3 - 4 | 625 | 68 | ||
| AK169449.1 | - | 3 - 4 | 626 | 68 | ||
| AK169957.1 | - | 3 - 4 | 663 | 68 | ||
| BC055804.1 | - | 3 - 4 | 605 | 68 | ||
| BC094608.1 | - | 4163 | 68 | |||
| BC139371.1 | - | 3 - 4 | 626 | 8862393 | 68 | |
| BC139397.1 | - | 3 - 4 | 626 | 8862419 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef4
Genomic Map