Assay Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290801.1 | NP_001277730.1 | 1 - 2 | 359 | 113 | |
| NM_173870.3 | NP_776295.1 | 2 - 3 | 411 | 113 | ||
| GenBank mRNA | AB053217.1 | - | 1 - 2 | 87 | 113 | |
| AK155724.1 | - | 2 - 3 | 412 | 113 | ||
| AK163327.1 | - | 1 - 2 | 191 | 113 | ||
More Information
Set Membership: |
|

Mgat4a
Genomic Map