Assay Details
| Gene Symbol: Cnnm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033570.2 | NP_291048.2 | 1 - 2 | 1484 | 79 | |
| GenBank mRNA | AF216963.1 | - | 1 - 2 | 1096 | 79 | |
| AK173209.1 | - | 1 - 2 | 1461 | 79 | ||
| GU390541.1 | - | 1 - 2 | 1019 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm4
Genomic Map