Assay Details
| Gene Symbol: Cnnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039551.1 | NP_001034640.1 | 1 - 2 | 1280 | 71 | |
| NM_053186.2 | NP_444416.2 | 1 - 2 | 1280 | 71 | ||
| GenBank mRNA | AF216964.1 | - | 1 - 2 | 349 | 71 | |
| BC052714.1 | - | 1 - 2 | 350 | 71 | ||
| BC108417.1 | - | 1 - 2 | 534 | 71 | ||
| BC132285.1 | - | 1 - 2 | 332 | 71 | ||
| BC144720.1 | - | 1 - 2 | 331 | 71 | ||
| GQ225746.2 | - | 1 - 2 | 266 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm3
Genomic Map