Assay Details
| Gene Symbol: Msh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010830.2 | NP_034960.1 | 5 - 6 | 3529 | 81 | |
| GenBank mRNA | AK010643.1 | - | 2 - 3 | 690 | 81 | |
| AK088259.1 | - | 5 - 6 | 3518 | 81 | ||
| BC035274.1 | - | 3 - 4 | 2927 | 81 | ||
| BC051160.1 | - | 5 - 6 | 3474 | 4953238 | 81 | |
| BC051634.1 | - | 5 - 6 | 3484 | 6518577 | 81 | |
| U42190.1 | - | 5 - 6 | 3505 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh6
Genomic Map