Assay Details
| Gene Symbol: Msh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010830.2 | NP_034960.1 | 7 - 8 | 3741 | 74 | |
| GenBank mRNA | AK010643.1 | - | 4 - 5 | 902 | 74 | |
| AK088259.1 | - | 7 - 8 | 3730 | 74 | ||
| BC035274.1 | - | 5 - 6 | 3139 | 74 | ||
| BC051160.1 | - | 7 - 8 | 3686 | 4953238 | 74 | |
| BC051634.1 | - | 7 - 8 | 3696 | 6518577 | 74 | |
| U42190.1 | - | 7 - 8 | 3717 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh6
Genomic Map