Assay Details
| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163300.1 | NP_001156772.1 | 10 - 11 | 1689 | 68 | |
| GenBank mRNA | AK087504.1 | - | 3 - 4 | 2149 | 68 | |
More Information
Set Membership: |
|

Safb
Genomic Map