Assay Details
| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163300.1 | NP_001156772.1 | 12 - 13 | 1913 | 52 | |
| GenBank mRNA | AK087504.1 | - | 5 - 6 | 2373 | 52 | |
More Information
Set Membership: |
|

Safb
Genomic Map