Assay Details
| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163300.1 | NP_001156772.1 | 13 - 14 | 2000 | 79 | |
| GenBank mRNA | AK087504.1 | - | 6 - 7 | 2460 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Safb
Genomic Map