Assay Details
| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163300.1 | NP_001156772.1 | 14 - 15 | 2104 | 95 | |
| GenBank mRNA | AK087504.1 | - | 7 - 8 | 2564 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Safb
Genomic Map