Assay Details
| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163300.1 | NP_001156772.1 | 4 - 5 | 724 | 96 | |
| GenBank mRNA | BC018200.1 | - | 4 - 5 | 700 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Safb
Genomic Map