Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 11 - 12 | 2618 | 69 | |
| GenBank mRNA | AF064984.1 | - | 11 - 12 | 2608 | 69 | |
| AF077847.1 | - | 11 - 12 | 2496 | 69 | ||
| AK090382.1 | - | 2 - 3 | 324 | 69 | ||
| AK147068.1 | - | 11 - 12 | 2611 | 69 | ||
| AK147231.1 | - | 12 - 13 | 2729 | 69 | ||
| AK147330.1 | - | 12 - 13 | 2729 | 69 | ||
| AK220186.1 | - | 11 - 12 | 2571 | 69 | ||
| BC011374.1 | - | 11 - 12 | 2600 | 3154246 | 69 | |
| BC094610.1 | - | 11 - 12 | 2607 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp5
Genomic Map