Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 13 - 14 | 3141 | 75 | |
| GenBank mRNA | AF064984.1 | - | 13 - 14 | 3131 | 75 | |
| AF077847.1 | - | 13 - 14 | 3019 | 75 | ||
| AK090382.1 | - | 4 - 5 | 819 | 75 | ||
| AK147068.1 | - | 13 - 14 | 3134 | 75 | ||
| AK147231.1 | - | 14 - 15 | 3252 | 75 | ||
| AK147330.1 | - | 14 - 15 | 3252 | 75 | ||
| AK220186.1 | - | 13 - 14 | 3094 | 75 | ||
| BC011374.1 | - | 13 - 14 | 3123 | 3154246 | 75 | |
| BC094610.1 | - | 13 - 14 | 3129 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp5
Genomic Map