Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 1 - 2 | 211 | 61 | |
| GenBank mRNA | AF064984.1 | - | 1 - 2 | 201 | 61 | |
| AF077847.1 | - | 1 - 2 | 89 | 61 | ||
| AK081025.1 | - | 1 - 2 | 211 | 61 | ||
| AK147068.1 | - | 1 - 2 | 204 | 61 | ||
| AK147231.1 | - | 1 - 2 | 209 | 61 | ||
| AK147330.1 | - | 1 - 2 | 209 | 61 | ||
| AK220186.1 | - | 1 - 2 | 164 | 61 | ||
| BC011374.1 | - | 1 - 2 | 193 | 3154246 | 61 | |
| BC094610.1 | - | 1 - 2 | 200 | 61 | ||
More Information
Set Membership: |
|

Lrp5
Genomic Map