Assay Details
| Gene Symbol: Fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081034.1 | NP_001074503.1 | 8 - 9 | 1084 | 75 | |
| GenBank mRNA | BC110686.1 | - | 8 - 9 | 823 | 75 | |
| BC117884.1 | - | 8 - 9 | 830 | 40096054 | 75 | |
| BC117885.1 | - | 8 - 9 | 834 | 40096055 | 75 | |
| BC128479.1 | - | 8 - 9 | 834 | 40096053 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo11
Genomic Map