Assay Details
| Gene Symbol: Spic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011461.3 | NP_035591.3 | 2 - 3 | 143 | 85 | |
| GenBank mRNA | AF098863.2 | - | 2 - 3 | 142 | 85 | |
| AK041260.1 | - | 2 - 3 | 144 | 85 | ||
| AK137522.1 | - | 2 - 3 | 146 | 85 | ||
| AK156024.1 | - | 2 - 3 | 144 | 85 | ||
| AK156892.1 | - | 2 - 3 | 144 | 85 | ||
| AK157188.1 | - | 2 - 3 | 145 | 85 | ||
| BC064038.1 | - | 1 - 2 | 104 | 3824951 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF098863.2, AK137522.1, AK156024.1, AK156892.1, AK157188.1, BC064038.1, NM_011461.3 transcripts.
Set Membership: |
|

Spic
Genomic Map