Assay Details
| Gene Symbol: Ccdc88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081291.1 | NP_001074760.1 | 23 - 24 | 4026 | 82 | |
| GenBank mRNA | BC076600.1 | - | 14 - 15 | 2910 | 82 | |
| BC151001.1 | - | 23 - 24 | 3999 | 9087914 | 82 | |
| BC151009.1 | - | 23 - 24 | 3999 | 9087922 | 82 | |
| BC157946.1 | - | 23 - 24 | 3999 | 9088122 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc88b
Genomic Map