Assay Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175930.5 | NP_787126.3 | 17 - 18 | 1990 | 89 | |
| GenBank mRNA | AK029995.1 | - | 17 - 18 | 1991 | 89 | |
| AK030016.1 | - | 17 - 18 | 1944 | 89 | ||
| AK083591.1 | - | 7 - 8 | 930 | 89 | ||
| AK122234.1 | - | 5 - 6 | 752 | 89 | ||
| BC046627.1 | - | 5 - 6 | 711 | 6494175 | 89 | |
More Information
Set Membership: |
|

Rapgef5
Genomic Map