Assay Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175930.5 | NP_787126.3 | 22 - 23 | 2623 | 82 | |
| GenBank mRNA | AK030016.1 | - | 22 - 23 | 2577 | 82 | |
| AK083591.1 | - | 12 - 13 | 1563 | 82 | ||
| AK122234.1 | - | 10 - 11 | 1385 | 82 | ||
| BC046627.1 | - | 10 - 11 | 1344 | 6494175 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rapgef5
Genomic Map