Assay Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175930.5 | NP_787126.3 | 24 - 25 | 2760 | 100 | |
| GenBank mRNA | AK029995.1 | - | 24 - 25 | 2761 | 100 | |
| AK030016.1 | - | 24 - 25 | 2714 | 100 | ||
| AK083591.1 | - | 14 - 15 | 1700 | 100 | ||
| AK122234.1 | - | 12 - 13 | 1522 | 100 | ||
| BC046627.1 | - | 12 - 13 | 1481 | 6494175 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rapgef5
Genomic Map