Assay Details
| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008073.3 | NP_032099.1 | 2 - 3 | 640 | 91 | |
| NM_177408.6 | NP_803127.3 | 2 - 3 | 640 | 91 | ||
| GenBank mRNA | AF233775.1 | - | 2 - 3 | 256 | 91 | |
| AF233777.1 | - | 2 - 3 | 256 | 91 | ||
| AF233782.1 | - | 2 - 3 | 256 | 91 | ||
| AF233784.1 | - | 2 - 3 | 256 | 91 | ||
| AF233787.1 | - | 2 - 3 | 256 | 91 | ||
| AF233788.1 | - | 2 - 3 | 256 | 91 | ||
| AF233789.1 | - | 2 - 3 | 256 | 91 | ||
| AF233790.1 | - | 2 - 3 | 256 | 91 | ||
| AF233791.1 | - | 2 - 3 | 256 | 91 | ||
| AF233795.1 | - | 2 - 3 | 256 | 91 | ||
| AF233798.1 | - | 2 - 3 | 256 | 91 | ||
| AF233800.1 | - | 2 - 3 | 256 | 91 | ||
| AF233801.1 | - | 2 - 3 | 256 | 91 | ||
| AK034338.1 | - | 2 - 3 | 610 | 91 | ||
| AK047085.1 | - | 2 - 3 | 218 | 91 | ||
| AK049857.1 | - | 2 - 3 | 283 | 91 | ||
| AK136889.1 | - | 2 - 3 | 248 | 91 | ||
| AK141129.1 | - | 2 - 3 | 282 | 91 | ||
| BC031762.1 | - | 2 - 3 | 458 | 4504205 | 91 | |
| M62374.1 | - | 2 - 3 | 603 | 91 | ||
More Information
Set Membership: |
|

Gabrg2
Genomic Map