Assay Details
| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008073.3 | NP_032099.1 | 8 - 9 | 1512 | 100 | |
| GenBank mRNA | AF233775.1 | - | 8 - 9 | 1128 | 100 | |
| AF233776.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233777.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233778.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233779.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233780.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233781.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233782.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233783.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233784.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233785.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233786.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233787.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233788.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233789.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233790.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233791.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233792.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233793.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233794.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233795.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233796.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233797.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233798.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233799.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233800.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233801.1 | - | 8 - 9 | 1128 | 100 | ||
| AF233802.1 | - | 8 - 9 | 1128 | 100 | ||
| AK047085.1 | - | 8 - 9 | 1090 | 100 | ||
| BC031762.1 | - | 8 - 9 | 1330 | 4504205 | 100 | |
| M62374.1 | - | 8 - 9 | 1475 | 100 | ||
| X56313.1 | - | 1 - 2 | 204 | 100 | ||
More Information
Set Membership: |
|

Gabrg2
Genomic Map