Assay Details
| Gene Symbol: Ganab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293621.1 | NP_001280550.1 | 1 - 2 | 104 | 78 | |
| NM_008060.2 | NP_032086.1 | 1 - 2 | 104 | 78 | ||
| GenBank mRNA | AK028655.1 | - | 1 - 2 | 72 | 78 | |
| AK030722.1 | - | 1 - 2 | 73 | 78 | ||
| AK043606.1 | - | 1 - 2 | 51 | 78 | ||
| AK081915.1 | - | 1 - 2 | 66 | 78 | ||
| AK149686.1 | - | 1 - 2 | 61 | 78 | ||
| BC094437.1 | - | 1 - 2 | 54 | 30682580 | 78 | |
| BC117888.1 | - | 1 - 2 | 47 | 40096413 | 78 | |
| BC117889.1 | - | 1 - 2 | 50 | 40096414 | 78 | |
| BC128069.1 | - | 1 - 2 | 48 | 40096428 | 78 | |
| U92793.1 | - | 1 - 2 | 50 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030722.1, AK081915.1, AK149686.1, BC094437.1, BC117888.1, BC117889.1, BC128069.1, NM_001293621.1, NM_008060.2, U92793.1 transcripts.
Set Membership: |
|

Ganab
Genomic Map