Assay Details
| Gene Symbol: Tut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197993.2 | NP_932110.1 | 4 - 5 | 777 | 63 | |
| GenBank mRNA | AK138418.1 | - | 4 - 5 | 697 | 63 | |
| BC023900.1 | - | 4 - 5 | 754 | 5321265 | 63 | |
| BC025499.1 | - | 4 - 5 | 801 | 5320398 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tut1
Genomic Map