Assay Details
| Gene Symbol: Tut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197993.2 | NP_932110.1 | 6 - 7 | 1371 | 120 | |
| GenBank mRNA | BC023900.1 | - | 6 - 7 | 1348 | 5321265 | 120 |
| BC025499.1 | - | 6 - 7 | 1395 | 5320398 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tut1
Genomic Map