Assay Details
| Gene Symbol: Tut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197993.2 | NP_932110.1 | 8 - 9 | 1571 | 65 | |
| GenBank mRNA | BC023900.1 | - | 8 - 9 | 1548 | 5321265 | 65 |
| BC025499.1 | - | 8 - 9 | 1595 | 5320398 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA polyadenylation
mRNA cleavage
mRNA processing
snRNA processing
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding
polynucleotide adenylyltransferase activity
ATP binding
transferase activity
nucleotidyltransferase activity
enzyme binding
metal ion binding
RNA uridylyltransferase activity
mRNA polyadenylation
mRNA cleavage
mRNA processing
snRNA processing
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding
polynucleotide adenylyltransferase activity
ATP binding
transferase activity
nucleotidyltransferase activity
enzyme binding
metal ion binding
RNA uridylyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986199 | BC023900.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986199 | BC025499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986199 | AK215597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986199 | NM_197993.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151392 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121964 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173746 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986199 | BC023900.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986199 | BC025499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986199 | AK215597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986199 | NM_197993.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433556 | BC025499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433556 | NM_197993.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134136 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123892 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14418063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14342711 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14342728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14342721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14342731 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14342734 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14342729 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461358 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461359 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440395_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451106_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461354 | - | - |
| Mouse Genome 430 2.0 Array | 1451106_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440395_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451106_at | - | - |
| Mouse Genome 430 2.0 Array | 1440395_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70044_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70044_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900002319.mm.1 | NM_197993.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002319.mm.1 | TC1900000169.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451106_at | - | - |
| Mouse Expression Set 430 | 1451106_PM_at | - | - |
| Mouse Expression Set 430 | 1440395_PM_at | - | - |
| Mouse Expression Set 430 | 1440395_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451106_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451106_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451106_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451106_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440395_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440395_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 302285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6350309 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105340671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232593 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232593 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215298 | - | - |
| mouse-6 v1.1 (Illumina) | scl53393.10_649-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350309 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105340671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53393.10_649-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350309 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105340671 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232593 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232593 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215298 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53393.10_649-S | - | - |

Tut1
Genomic Map