Assay Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026720.2 | NP_080996.2 | 12 - 13 | 1515 | 77 | |
| GenBank mRNA | AK002981.1 | - | 1 - 2 | 41 | 77 | |
| AK089186.1 | - | 12 - 13 | 1508 | 77 | ||
| BC058982.1 | - | 12 - 13 | 1426 | 5685059 | 77 | |
More Information
Set Membership: |
|

Ankrd13d
Genomic Map