Assay Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026720.2 | NP_080996.2 | 13 - 14 | 1681 | 66 | |
| GenBank mRNA | AK002981.1 | - | 2 - 3 | 205 | 66 | |
| AK089186.1 | - | 13 - 14 | 1674 | 66 | ||
| AK207732.1 | - | 1 - 2 | 105 | 66 | ||
| BC058982.1 | - | 13 - 14 | 1592 | 5685059 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922243 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170978 | BC058982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170978 | NM_026720.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170978 | AK089186.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73377 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124714 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174351 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170978 | BC058982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170978 | NM_026720.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170978 | AK089186.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31922235 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174351 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124714 | - | - |
| Whole Mouse Genome G4122A | A_51_P511029 | BC058982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511029 | NM_026720.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511029 | AK089186.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140833 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114356 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124714 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436752 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436754 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31922242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14498647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14407457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14428562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14142413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14347411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14164640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14196711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14407468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14407464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14428564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14286977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14237252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14454243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10464697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464693 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464690 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452965_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447738_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10464688 | - | - |
| Mouse Genome 430 2.0 Array | 1447738_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452965_at | - | - |
| Mouse Genome 430 2.0 Array | 1452965_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447738_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68423_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68423_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900007961.mm.1 | NM_026720.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007961.mm.1 | NM_026720.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007961.mm.1 | TC1900000915.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452965_PM_at | - | - |
| Mouse Expression Set 430 | 1452965_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452965_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447738_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452965_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447738_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4280164 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700477 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2700477 | - | - |
| mouse-6 v1.1 (Illumina) | scl53451.16.1_293-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53451.16.1_293-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2851735 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2851735 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53451.16.1_293-S | - | - |

Ankrd13d
Genomic Map