Assay Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026720.2 | NP_080996.2 | 1 - 2 | 262 | 101 | |
| GenBank mRNA | AK089186.1 | - | 1 - 2 | 263 | 101 | |
| AK155698.1 | - | 1 - 2 | 263 | 101 | ||
| BC058982.1 | - | 1 - 2 | 173 | 5685059 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058982.1 transcripts.
Set Membership: |
|

Ankrd13d
Genomic Map