Assay Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026720.2 | NP_080996.2 | 4 - 5 | 565 | 72 | |
| GenBank mRNA | AK089186.1 | - | 4 - 5 | 558 | 72 | |
| AK155698.1 | - | 4 - 5 | 566 | 72 | ||
| BC058982.1 | - | 4 - 5 | 476 | 5685059 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13d
Genomic Map