Assay Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026720.2 | NP_080996.2 | 5 - 6 | 708 | 72 | |
| GenBank mRNA | AK089186.1 | - | 5 - 6 | 701 | 72 | |
| AK155698.1 | - | 5 - 6 | 709 | 72 | ||
| BC058982.1 | - | 5 - 6 | 619 | 5685059 | 72 | |
More Information
Set Membership: |
|

Ankrd13d
Genomic Map