Assay Details
| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198650.2 | NP_941052.1 | 1 - 2 | 381 | 59 | |
| GenBank mRNA | AY194118.1 | - | 1 - 2 | 380 | 59 | |
| BC046588.1 | - | 1 - 2 | 381 | 6309674 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a20
Genomic Map