Assay Details
| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198650.2 | NP_941052.1 | 2 - 3 | 483 | 72 | |
| GenBank mRNA | AY194118.1 | - | 2 - 3 | 482 | 72 | |
| BC046588.1 | - | 2 - 3 | 483 | 6309674 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a20
Genomic Map