Assay Details
| Gene Symbol: Arhgap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039692.1 | NP_001034781.1 | 11 - 12 | 2022 | 142 | |
| NM_029277.2 | NP_083553.2 | 10 - 11 | 1947 | 142 | ||
| GenBank mRNA | AK031648.1 | - | 11 - 12 | 1866 | 142 | |
| BC024633.1 | - | 6 - 7 | 434 | 142 | ||
| BC037608.1 | - | 10 - 11 | 1762 | 142 | ||
| BC141184.1 | - | 10 - 11 | 1756 | 9055796 | 142 | |
| BC145506.1 | - | 10 - 11 | 1755 | 9054069 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap12
Genomic Map