Assay Details
| Gene Symbol: Amt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013814.1 | NP_001013836.1 | 3 - 4 | 358 | 66 | |
| GenBank mRNA | BC132277.1 | - | 3 - 4 | 410 | 40130554 | 66 |
| BC132279.1 | - | 3 - 4 | 410 | 40130556 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amt
Genomic Map