Assay Details
| Gene Symbol: Amt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013814.1 | NP_001013836.1 | 6 - 7 | 703 | 64 | |
| GenBank mRNA | BC132277.1 | - | 6 - 7 | 755 | 40130554 | 64 |
| BC132279.1 | - | 6 - 7 | 755 | 40130556 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975052 | NM_001013814.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736086 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975052 | NM_001013814.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814153 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06919946 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907265 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07613160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07964226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07659984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07907254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07907261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07907266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07964227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26814157 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588892 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588883 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900011229.mm.1 | NM_001013814.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011229.mm.1 | NM_001013814.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011229.mm.1 | TC0900001399.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160992 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160992 | - | - |
| mouse-6 v1.1 (Illumina) | GI_62000669-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160992 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160992 | - | - |

Amt
Genomic Map