Assay Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011678.2 | NP_035808.2 | 10 - 11 | 1377 | 61 | |
| GenBank mRNA | AK143582.1 | - | 10 - 11 | 1312 | 61 | |
| AK149964.1 | - | 10 - 11 | 1335 | 61 | ||
| AK164245.1 | - | 1 - 2 | 152 | 61 | ||
| AK169372.1 | - | 10 - 11 | 1356 | 61 | ||
| AK169933.1 | - | 10 - 11 | 1347 | 61 | ||
| AK171271.1 | - | 10 - 11 | 1347 | 61 | ||
| AK171633.1 | - | 10 - 11 | 1330 | 61 | ||
| AK220512.1 | - | 10 - 11 | 1334 | 61 | ||
| BC011341.1 | - | 9 - 10 | 1154 | 3661165 | 61 | |
| BC066180.1 | - | 10 - 11 | 1287 | 61 | ||
| BC066865.1 | - | 10 - 11 | 1287 | 61 | ||
| L00681.1 | - | 10 - 11 | 1340 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp4
Genomic Map