Assay Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011678.2 | NP_035808.2 | 17 - 18 | 2351 | 63 | |
| GenBank mRNA | AK143582.1 | - | 17 - 18 | 2286 | 63 | |
| AK149964.1 | - | 17 - 18 | 2309 | 63 | ||
| AK164245.1 | - | 8 - 9 | 1126 | 63 | ||
| AK169372.1 | - | 18 - 19 | 2330 | 63 | ||
| AK169933.1 | - | 17 - 18 | 2321 | 63 | ||
| AK171271.1 | - | 17 - 18 | 2321 | 63 | ||
| AK171633.1 | - | 17 - 18 | 2304 | 63 | ||
| AK181598.1 | - | 3 - 4 | 384 | 63 | ||
| AK206204.1 | - | 2 - 3 | 304 | 63 | ||
| AK214933.1 | - | 3 - 4 | 379 | 63 | ||
| BC011341.1 | - | 16 - 17 | 2128 | 3661165 | 63 | |
| BC066180.1 | - | 17 - 18 | 2261 | 63 | ||
| BC066865.1 | - | 17 - 18 | 2261 | 63 | ||
| L00681.1 | - | 17 - 18 | 2314 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp4
Genomic Map