Assay Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011678.2 | NP_035808.2 | 7 - 8 | 928 | 62 | |
| GenBank mRNA | AK143582.1 | - | 7 - 8 | 863 | 62 | |
| AK149964.1 | - | 7 - 8 | 886 | 62 | ||
| AK169372.1 | - | 7 - 8 | 907 | 62 | ||
| AK169933.1 | - | 7 - 8 | 898 | 62 | ||
| AK171271.1 | - | 7 - 8 | 898 | 62 | ||
| AK171633.1 | - | 7 - 8 | 881 | 62 | ||
| AK220512.1 | - | 7 - 8 | 885 | 62 | ||
| BC066180.1 | - | 7 - 8 | 838 | 62 | ||
| BC066865.1 | - | 7 - 8 | 838 | 62 | ||
More Information
Set Membership: |
|

Usp4
Genomic Map