Assay Details
| Gene Symbol: Gm4850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015347.1 | 1 - 2 | 140 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PIH1 domain containing 3 | Pih1d3 | 4930521A18Rik | -40.6kb | Sense | Chr. 1 - 31222839 - 31224287 on NCBI Build GRCm38 |
| Downstream |
KH domain containing, RNA binding, signal transduction associated 2 |
Khdrbs2 |
6330586C16Rik, Slim1, Slm1 |
904.6kb | Sense | Chr. 1 - 32172688 - 32705224 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4850
Genomic Map